This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TSC22D1 SIRNA Assays ›
Gene Symbol:
Tsc22d1
Gene Name:
TSC22 domain family, member 1
Gene Aliases:
TS22A, TSC-22, Tgfb1i4
Chromosome Location:
Chr.15: 58554358 - 58658156 on Build Rnor_6.0
UniGene ID:
Rn.3545
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 49006

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tsc22d1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109912.1 NP_001103382.1 3 3279
NM_013043.2 NP_037175.1 3 407
XM_006252339.1 XP_006252401.1 4 3151
GenBank mRNA BC059146.1 - 3 418
BC127547.1 - 3 399
L25785.1 - 3 400

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation
positive regulation of apoptotic process
negative regulation of apoptotic process
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-976xq:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline