siRNA Details
| Gene Symbol: Nqo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017000.3 | NP_058696.2 | 2 | 197 | ||
| GenBank mRNA | BC083542.1 | - | 2 | 197 | 7190779 | |
| J02608.1 | - | 2 | 182 | |||
| J02640.1 | - | 1 | 156 | |||
| J02679.1 | - | 2 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nqo1| Gene Symbol: Nqo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017000.3 | NP_058696.2 | 2 | 197 | ||
| GenBank mRNA | BC083542.1 | - | 2 | 197 | 7190779 | |
| J02608.1 | - | 2 | 182 | |||
| J02640.1 | - | 1 | 156 | |||
| J02679.1 | - | 2 | 182 | |||