siRNA Details
| Gene Symbol: Grm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017012.1 | NP_058708.1 | 2 | 508 | ||
| XM_006229659.2 | XP_006229721.1 | 2 | 835 | |||
| GenBank mRNA | D10891.1 | - | 2 | 508 | ||
| EU559292.1 | - | 1 | 241 | |||
More Information
Set Membership: |
|

Grm5
Genomic Map