siRNA Details
| Gene Symbol: Il1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017019.1 | NP_058715.1 | 4 | 372 | ||
| GenBank mRNA | AJ245642.1 | - | 4 | 336 | ||
| AJ245643.1 | - | 3 | 311 | |||
| D00403.1 | - | 4 | 372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1a| Gene Symbol: Il1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017019.1 | NP_058715.1 | 4 | 372 | ||
| GenBank mRNA | AJ245642.1 | - | 4 | 336 | ||
| AJ245643.1 | - | 3 | 311 | |||
| D00403.1 | - | 4 | 372 | |||