siRNA Details
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017094.1 | NP_058790.1 | 4 | 429 | ||
XM_008760752.1 | XP_008758974.1 | 4 | 483 | |||
XM_008760753.1 | XP_008758975.1 | 4 | 364 | |||
XM_008760754.1 | XP_008758976.1 | 4 | 707 | |||
GenBank mRNA | BC086591.1 | - | 4 | 412 | ||
BC103659.1 | - | 4 | 391 | |||
BC127501.1 | - | 4 | 470 | |||
BC161852.1 | - | 4 | 416 | |||
FQ216498.1 | - | 4 | 485 | |||
FQ219220.1 | - | 4 | 461 | |||
J04811.1 | - | 4 | 429 | |||
JF330099.1 | - | 2 | 169 | |||
JF412704.1 | - | 3 | 239 | |||
S49003.1 | - | 4 | 429 |
