This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ESP18 SIRNA Assays ›
Gene Symbol:
Esp18
Gene Name:
exocrine gland secreted peptide 18
Gene Aliases:
Gm20579
Chromosome Location:
Chr.17: 39406359 - 39410992 on Build GRCm38
UniGene ID:
Mm.350534
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 500356

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Esp18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244763.1 NP_001231692.1 3 612
GenBank mRNA AB306997.1 - 4 590

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
pheromone activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-7zhdl:80/100.66.77.60:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0