This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PAK1 SIRNA Assays ›
Gene Symbol:
Pak1
Gene Name:
p21 protein (Cdc42/Rac)-activated kinase 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 162768156 - 162883356 on Build Rnor_6.0
UniGene ID:
Rn.9149
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 50059

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pak1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017198.1 NP_058894.1 7 1146
XM_006229752.1 XP_006229814.1 7 997
XM_006229753.1 XP_006229815.1 7 994
XM_006229754.2 XP_006229816.1 7 986
GenBank mRNA U23443.1 - 7 952
U49953.1 - 7 1146

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011035, BC145698, AF082077, AK077497.

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPKK activity
mitotic cell cycle
response to hypoxia
positive regulation of protein phosphorylation
protein phosphorylation
exocytosis
apoptotic process
small GTPase mediated signal transduction
axonogenesis
neuromuscular junction development
response to organic substance
dendrite development
actin cytoskeleton reorganization
cellular response to insulin stimulus
regulation of actin cytoskeleton organization
positive regulation of peptidyl-serine phosphorylation
positive regulation of intracellular estrogen receptor signaling pathway
intracellular signal transduction
wound healing
receptor clustering
positive regulation of JUN kinase activity
protein autophosphorylation
branching morphogenesis of an epithelial tube
neuron projection morphogenesis
positive regulation of stress fiber assembly
negative regulation of cell proliferation involved in contact inhibition
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
collagen binding
ATP binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline