siRNA Details
| Gene Symbol: Gm20172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204913.1 | NP_001191842.1 | 1 | 1088 | ||
| GenBank mRNA | AK084043.1 | - | 1 | 1088 | ||
| AK148788.1 | - | 1 | 1088 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20172| Gene Symbol: Gm20172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204913.1 | NP_001191842.1 | 1 | 1088 | ||
| GenBank mRNA | AK084043.1 | - | 1 | 1088 | ||
| AK148788.1 | - | 1 | 1088 | |||