siRNA Details
| Gene Symbol: Odf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145005.1 | NP_001138477.1 | 12, 13 | 1446 | ||
| NM_017213.2 | NP_058909.2 | 12, 13 | 1572 | |||
| GenBank mRNA | AF053971.1 | - | 11, 12 | 1673 | ||
| AF162755.1 | - | 8, 9 | 966 | |||
| AF162756.2 | - | 1513 | ||||
| BC078857.1 | - | 12, 13 | 1572 | 7112637 | ||
| EF197115.1 | - | 1, 2 | 76 | |||
| U62821.1 | - | 10, 11 | 1404 | |||
| X95272.1 | - | 12, 13 | 1413 | |||

Odf2
Genomic Map