This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LAMA5 SIRNA Assays ›
Gene Symbol:
Lama5
Gene Name:
laminin, alpha 5
Gene Aliases:
AA408760, AA408762, AI853660, [a]5, laminin-511, mKIAA0533
Chromosome Location:
Chr.2: 180176373 - 180225905 on Build GRCm38
UniGene ID:
Mm.4339
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 501134

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lama5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081171.2 NP_001074640.1 71 9929
GenBank mRNA AK129157.1 - 32 4703
AK158204.1 - 4 556
BC020313.2 - 8 1101
BC043478.1 - 1 75
BC062207.1 - 9 1362
U37501.1 - 71 9623

Back To Top

More Information


Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
morphogenesis of a polarized epithelium
neural crest cell migration
hair follicle development
cell adhesion
integrin-mediated signaling pathway
muscle organ development
organ morphogenesis
morphogenesis of embryonic epithelium
cell migration
regulation of cell adhesion
lung development
regulation of cell migration
substrate adhesion-dependent cell spreading
regulation of cell proliferation
cilium assembly
odontogenesis of dentin-containing tooth
regulation of embryonic development
branching morphogenesis of an epithelial tube
branching involved in salivary gland morphogenesis
establishment of protein localization to plasma membrane
receptor binding
integrin binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline