This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CNKSR1 SIRNA Assays ›
Gene Symbol:
Cnksr1
Gene Name:
connector enhancer of kinase suppressor of Ras 1
Gene Aliases:
BC037758
Chromosome Location:
Chr.4: 134228041 - 134238416 on Build GRCm38
UniGene ID:
Mm.331958
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 501246

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnksr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081047.1 NP_001074516.1 21 1993
GenBank mRNA AK145910.1 - 21 1955
BC037758.1 - 2 223
BC113805.1 - 21 1958 40062208
BC113956.1 - 21 1958 40062209

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

Ras protein signal transduction
Rho protein signal transduction
protein binding, bridging

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-976xq:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline