siRNA Details
| Gene Symbol: Lclat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081071.2 | NP_001074540.1 | 1, 2 | 201 | ||
| NM_001177967.1 | NP_001171438.1 | 1, 2 | 180 | |||
| GenBank mRNA | AK144580.1 | - | 1, 2 | 144 | ||
| BC147499.1 | - | 1, 2 | 119 | 9056361 | ||
| BC158076.1 | - | 1, 2 | 140 | 9088168 | ||
| BC158079.1 | - | 1, 2 | 140 | 9088171 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008764537, XM_008764538.

Lclat1
Genomic Map