siRNA Details
| Gene Symbol: Psme2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017257.2 | NP_058953.1 | 3, 4 | 157 | ||
| GenBank mRNA | BC058486.1 | - | 3, 4 | 195 | 6918985 | |
| BC097331.1 | - | 3, 4 | 157 | 7389180 | ||
| D45250.1 | - | 3, 4 | 207 | |||
| FQ226805.1 | - | 3, 4 | 188 | |||
| FQ227049.1 | - | 3, 4 | 201 | |||
| FQ232840.1 | - | 3, 4 | 188 | |||
| FQ233763.1 | - | 3, 4 | 183 | |||

Psme2
Genomic Map