This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TNKS1BP1 SIRNA Assays ›
Gene Symbol:
Tnks1bp1
Gene Name:
tankyrase 1 binding protein 1
Gene Aliases:
Tab182, mKIAA1741
Chromosome Location:
Chr.2: 85048012 - 85073048 on Build GRCm38
UniGene ID:
Mm.23606
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 502122

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tnks1bp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081260.2 NP_001074729.1 12 5590
GenBank mRNA AK081682.1 - 4 1944
AK129436.1 - 8 4283
BC024499.1 - 7 1488
BC025657.1 - 5 1069
BC025943.1 - 7 3039
BC058639.1 - 9 4734

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_033396, AK074113, BC046216, BC023622, AF441771, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline