siRNA Details
| Gene Symbol: Ffar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033316.2 | NP_001028488.1 | 2 | 1153 | ||
| GenBank mRNA | AK148616.1 | - | 2 | 1154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ffar3| Gene Symbol: Ffar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033316.2 | NP_001028488.1 | 2 | 1153 | ||
| GenBank mRNA | AK148616.1 | - | 2 | 1154 | ||