This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GUCY1A2 SIRNA Assays ›
Gene Symbol:
Gucy1a2
Gene Name:
guanylate cyclase 1, soluble, alpha 2
Gene Aliases:
6330407I18Rik, A230060L24Rik
Chromosome Location:
Chr.9: 3532349 - 3905791 on Build GRCm38
UniGene ID:
Mm.91240
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 502464

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gucy1a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033322.2 NP_001028494.1 8 11226
GenBank mRNA AK038764.1 - 1 2546
AK039640.1 - 1 153
AK083446.1 - 1 1303

Back To Top

More Information


Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
positive regulation of cGMP biosynthetic process
guanylate cyclase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline