This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM4944 SIRNA Assays ›
Gene Symbol:
Gm4944
Gene Name:
predicted gene 4944
Gene Aliases:
EG240038
Chromosome Location:
Chr.17: 22197265 - 22225659 on Build GRCm38
UniGene ID:
Mm.227945
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 502697

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm4944
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205095.1 NP_001192024.1 5 3250
GenBank mRNA AK136656.1 - 1 1360

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-555ccb4fbd-r4fm9:80/100.66.72.35:80.
git-commit: 75f0c7d4cb44686da2af39b21f2e6446a20282e2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-2025.12.99-1.0