This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM4951 SIRNA Assays ›
Gene Symbol:
Gm4951
Gene Name:
predicted gene 4951
Gene Aliases:
EG240327, Ifgga2
Chromosome Location:
Chr.18: 60211990 - 60248440 on Build GRCm38
UniGene ID:
Mm.357104
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 502716

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm4951
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033767.3 NP_001028939.1 3 316
GenBank mRNA AK149556.1 - 2 208
AK149583.1 - 3 314
BC139145.1 - 2 144 8862167
BC139147.1 - 2 144 8862169

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular response to interferon-beta
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-c8kzw:80/100.66.79.138:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline