This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NBEA SIRNA Assays ›
Gene Symbol:
Nbea
Gene Name:
neurobeachin
Gene Aliases:
Lyst2, mKIAA1544
Chromosome Location:
Chr.3: 55625195 - 56183701 on Build GRCm38
UniGene ID:
Mm.384353
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 503210

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nbea
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030595.1 NP_085098.1 18 2834
GenBank mRNA Y18276.1 - 18 2834

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008761027.

Gene Ontology Categories:

Function(s) Process(es)

protein targeting
post-Golgi vesicle-mediated transport
protein localization
endosomal transport
phospholipid binding
protein kinase binding
protein kinase A binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9wwlr:80/100.66.77.227:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline