This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DPCR1 SIRNA Assays ›
Gene Symbol:
Dpcr1
Gene Name:
diffuse panbronchiolitis critical region 1 (human)
Gene Aliases:
Gm630
Chromosome Location:
Chr.17: 35635755 - 35643695 on Build GRCm38
UniGene ID:
Mm.296301
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 503250

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dpcr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033366.3 NP_001028538.1 2 645
GenBank mRNA AK131887.1 - 2 647
AK164929.1 - 2 646
BC150798.1 - 2 638 9087709

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein targeting to mitochondrion
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline