siRNA Details
Gene Symbol: E030030I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254744.1 | NP_001241673.1 | 2 | 1394 | ||
GenBank mRNA | AK045260.1 | - | 2 | 944 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E030030I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254744.1 | NP_001241673.1 | 2 | 1394 | ||
GenBank mRNA | AK045260.1 | - | 2 | 944 |