siRNA Details
| Gene Symbol: Gm813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033404.2 | NP_001028576.1 | 4 | 613 | ||
| GenBank mRNA | AK136264.1 | - | 4 | 614 | ||
| BC145581.1 | - | 4 | 583 | |||
| BC145922.1 | - | 4 | 612 | 40131225 | ||
| BC145924.1 | - | 4 | 612 | 40131227 | ||

Gm813
Genomic Map