siRNA Details
| Gene Symbol: D630045J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194061.2 | NP_918950.2 | 2 | 2503 | ||
| GenBank mRNA | BC079889.1 | - | 1 | 1865 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630045J12Rik| Gene Symbol: D630045J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194061.2 | NP_918950.2 | 2 | 2503 | ||
| GenBank mRNA | BC079889.1 | - | 1 | 1865 | ||