This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SCGB1C1 SIRNA Assays ›
Gene Symbol:
Scgb1c1
Gene Name:
secretoglobin, family 1C, member 1
Gene Aliases:
Ryd5
Chromosome Location:
Chr.7: 140845565 - 140846769 on Build GRCm38
UniGene ID:
Mm.270960
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 503911

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scgb1c1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099742.1 NP_001093212.1 3 290
GenBank mRNA AA462242.1 - 3 263

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107561, X60661.

Panther Classification:

Molecular Function -

cytokine

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline