This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LSMEM1 SIRNA Assays ›
Gene Symbol:
Lsmem1
Gene Name:
leucine-rich single-pass membrane protein 1
Gene Aliases:
Gm889
Chromosome Location:
Chr.12: 40176386 - 40199315 on Build GRCm38
UniGene ID:
Mm.390463
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 503976

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lsmem1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033437.2 NP_001028609.1 4 640
GenBank mRNA AK134025.1 - 687
AK142184.1 - 4 641
BC147386.1 - 4 560 9056448
BC147387.1 - 4 560 9056444
BC172052.1 - 4 564

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline