siRNA Details
| Gene Symbol: Gm973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013771.2 | NP_001013793.2 | 13, 14 | 2025 | ||
| GenBank mRNA | BC072561.1 | - | 13, 14 | 2025 | 5696757 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm973| Gene Symbol: Gm973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013771.2 | NP_001013793.2 | 13, 14 | 2025 | ||
| GenBank mRNA | BC072561.1 | - | 13, 14 | 2025 | 5696757 | |