This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP408 SIRNA Assays ›
Gene Symbol:
Zfp408
Gene Name:
zinc finger protein 408
Gene Aliases:
Gm1011
Chromosome Location:
Chr.2: 91643168 - 91649897 on Build GRCm38
UniGene ID:
Mm.330401
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 504109

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp408
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033451.2 NP_001028623.1 4 3719
GenBank mRNA AK132515.1 - 4 3720

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-mjcrx:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0