siRNA Details
| Gene Symbol: Olfr403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207622.1 | NP_997505.1 | 1 | 34 | ||
| GenBank mRNA | BC104289.1 | - | 1 | 34 | 40042769 | |
| BC104290.1 | - | 1 | 34 | 40042770 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr403| Gene Symbol: Olfr403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207622.1 | NP_997505.1 | 1 | 34 | ||
| GenBank mRNA | BC104289.1 | - | 1 | 34 | 40042769 | |
| BC104290.1 | - | 1 | 34 | 40042770 | ||