siRNA Details
| Gene Symbol: Olfr681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207557.2 | NP_997440.2 | 1 | 904 | ||
| GenBank mRNA | BC104298.1 | - | 1 | 904 | 40043142 | |
| BC104299.1 | - | 1 | 904 | 40043148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr681| Gene Symbol: Olfr681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207557.2 | NP_997440.2 | 1 | 904 | ||
| GenBank mRNA | BC104298.1 | - | 1 | 904 | 40043142 | |
| BC104299.1 | - | 1 | 904 | 40043148 | ||