siRNA Details
| Gene Symbol: BC049352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198971.1 | NP_001185900.1 | 1, 2 | 233 | ||
| GenBank mRNA | BC049352.1 | - | 1, 2 | 233 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049352| Gene Symbol: BC049352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198971.1 | NP_001185900.1 | 1, 2 | 233 | ||
| GenBank mRNA | BC049352.1 | - | 1, 2 | 233 | ||