siRNA Details
| Gene Symbol: Mfsd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033488.2 | NP_001028660.2 | 14 | 2888 | ||
| GenBank mRNA | AK162620.1 | - | 12 | 2633 | ||
| AK171660.1 | - | 9 | 3252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd2b| Gene Symbol: Mfsd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033488.2 | NP_001028660.2 | 14 | 2888 | ||
| GenBank mRNA | AK162620.1 | - | 12 | 2633 | ||
| AK171660.1 | - | 9 | 3252 | |||