siRNA Details
| Gene Symbol: Gm5549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270430.1 | NP_001257359.1 | 1 | 14 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5549| Gene Symbol: Gm5549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270430.1 | NP_001257359.1 | 1 | 14 | ||