siRNA Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081069.1 | NP_001074538.1 | 8 | 565 | ||
| GenBank mRNA | AF061934.1 | - | 7 | 470 | ||
| AK039314.1 | - | 7 | 1181 | |||
| AK080707.1 | - | 8 | 655 | |||
| AK161551.1 | - | 4 | 618 | |||
| BC037655.1 | - | 6 | 1246 | |||
| BC049910.1 | - | 6 | 1263 | |||

Rgs11
Genomic Map