siRNA Details
| Gene Symbol: 1110037F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081183.1 | NP_001074652.1 | 10 | 2355 | ||
| GenBank mRNA | AK034232.1 | - | 9 | 2127 | ||
| AK052499.1 | - | 9 | 2143 | |||
| AK076731.1 | - | 9 | 2175 | |||
| AK122509.1 | - | 9 | 2108 | |||
| AK155623.1 | - | 9 | 2206 | |||
| AK170075.1 | - | 9 | 2208 | |||
| BC033309.1 | - | 10 | 2325 | 5024976 | ||

1110037F02Rik
Genomic Map