siRNA Details
| Gene Symbol: 1110037F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081183.1 | NP_001074652.1 | 9 | 2244 | ||
| GenBank mRNA | BC033309.1 | - | 9 | 2214 | 5024976 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110037F02Rik| Gene Symbol: 1110037F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081183.1 | NP_001074652.1 | 9 | 2244 | ||
| GenBank mRNA | BC033309.1 | - | 9 | 2214 | 5024976 | |