siRNA Details
| Gene Symbol: Gm8267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162954.1 | NP_001156426.1 | 4 | 675 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8267| Gene Symbol: Gm8267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162954.1 | NP_001156426.1 | 4 | 675 | ||