siRNA Details
| Gene Symbol: Tef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019194.2 | NP_062067.2 | 4 | 806 | ||
| XM_006242068.2 | XP_006242130.1 | 4 | 875 | |||
| GenBank mRNA | BC101895.1 | - | 4 | 806 | 7932583 | |
| S58745.1 | - | 4 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tef| Gene Symbol: Tef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019194.2 | NP_062067.2 | 4 | 806 | ||
| XM_006242068.2 | XP_006242130.1 | 4 | 875 | |||
| GenBank mRNA | BC101895.1 | - | 4 | 806 | 7932583 | |
| S58745.1 | - | 4 | 629 | |||