This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EFR3B SIRNA Assays ›
Gene Symbol:
Efr3b
Gene Name:
EFR3 homolog B (S. cerevisiae)
Gene Aliases:
AI852640, C030014M07Rik, mKIAA0953
Chromosome Location:
Chr.12: 3962554 - 4039094 on Build GRCm38
UniGene ID:
Mm.210266
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 506390

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Efr3b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082483.1 NP_001075952.1 15 1823
GenBank mRNA AK129247.1 - 11 1270
BC075650.1 - 14 1580
BC147643.1 - 15 1823 9056633
BC157968.1 - 15 1823 9088136

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-c7pcq:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0