siRNA Details
| Gene Symbol: Kif26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097621.1 | NP_001091090.1 | 16 | 6027 | ||
| GenBank mRNA | AK129315.1 | - | 8 | 4154 | ||
| BC059062.1 | - | 5 | 2858 | |||
| BC094358.1 | - | 5 | 3532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif26a| Gene Symbol: Kif26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097621.1 | NP_001091090.1 | 16 | 6027 | ||
| GenBank mRNA | AK129315.1 | - | 8 | 4154 | ||
| BC059062.1 | - | 5 | 2858 | |||
| BC094358.1 | - | 5 | 3532 | |||