siRNA Details
| Gene Symbol: Gm9573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244654.1 | NP_001231583.1 | 374 | |||
| GenBank mRNA | HM596785.1 | - | 376 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9573| Gene Symbol: Gm9573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244654.1 | NP_001231583.1 | 374 | |||
| GenBank mRNA | HM596785.1 | - | 376 | |||