siRNA Details
| Gene Symbol: 4930579F01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163385.1 | NP_001156857.1 | 1 | 171 | ||
| NM_001163386.1 | NP_001156858.1 | 1 | 171 | |||
| GenBank mRNA | AK016318.1 | - | 1 | 171 | ||
| BC150693.1 | - | 1 | 131 | 9087604 | ||

4930579F01Rik
Genomic Map