siRNA Details
| Gene Symbol: 1700012B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162428.1 | NP_001155900.1 | 6 | 1192 | ||
| GenBank mRNA | AK132924.1 | - | 1193 | |||
| AK160153.1 | - | 6 | 1194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700012B07Rik| Gene Symbol: 1700012B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162428.1 | NP_001155900.1 | 6 | 1192 | ||
| GenBank mRNA | AK132924.1 | - | 1193 | |||
| AK160153.1 | - | 6 | 1194 | |||