This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other R3HCC1 SIRNA Assays ›
Gene Symbol:
R3hcc1
Gene Name:
R3H domain and coiled-coil containing 1
Gene Aliases:
1700020M16Rik, AA407796
Chromosome Location:
Chr.14: 69697304 - 69714809 on Build GRCm38
UniGene ID:
Mm.27738
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 507660

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: R3hcc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146012.2 NP_001139484.2 3 266
NM_001301651.1 NP_001288580.1 3 218
NM_001301652.1 NP_001288581.1 2 499
GenBank mRNA AK006177.1 - 3 218
AK032854.1 - 3 236
AK156843.1 - 3 236
BC052536.1 - 2 415
BC059860.1 - 2 499
CJ181933.1 - 3 263

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
nucleic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline