siRNA Details
| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081381.1 | NP_001074850.1 | 6 | 651 | ||
| GenBank mRNA | AK011270.1 | - | 7 | 807 | ||
| AK075986.1 | - | 6 | 648 | |||
| BC048732.1 | - | 6 | 626 | 6512688 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elp6| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081381.1 | NP_001074850.1 | 6 | 651 | ||
| GenBank mRNA | AK011270.1 | - | 7 | 807 | ||
| AK075986.1 | - | 6 | 648 | |||
| BC048732.1 | - | 6 | 626 | 6512688 | ||