siRNA Details
| Gene Symbol: Fance | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163819.1 | NP_001157291.1 | 2 | 578 | ||
| NM_001163820.1 | NP_001157292.1 | 2 | 578 | |||
| GenBank mRNA | AK013325.1 | - | 2 | 541 | ||
| AK135017.1 | - | 3 | 482 | |||
| AK172283.1 | - | 2 | 562 | |||
| AK182639.1 | - | 1 | 236 | |||
| BC016538.1 | - | 2 | 536 | |||
| BC039987.1 | - | 2 | 541 | |||
| BY740269.1 | - | 2 | 562 | |||

Fance
Genomic Map