siRNA Details
| Gene Symbol: Tex33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163612.2 | NP_001157084.1 | 3 | 362 | ||
| NM_001163613.1 | NP_001157085.1 | 2 | 157 | |||
| NM_028522.1 | NP_082798.1 | 4 | 287 | |||
| GenBank mRNA | AK006856.1 | - | 4 | 261 | ||
| BC100472.1 | - | 3 | 335 | |||
| BC107197.2 | - | 3 | 166 | 40051085 | ||
| BC107198.1 | - | 3 | 166 | 40051089 | ||
| BY105066.1 | - | 3 | 352 | |||
| CJ047876.1 | - | 4 | 288 | |||

Tex33
Genomic Map