This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700125H20RIK SIRNA Assays ›
Gene Symbol:
1700125H20Rik
Gene Name:
RIKEN cDNA 1700125H20 gene
Gene Aliases:
-
Chromosome Location:
Chr.11: 85140088 - 85181164 on Build GRCm38
UniGene ID:
Mm.48850
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 508131

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700125H20Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028589.1 NP_082865.1 6 758
GenBank mRNA AK007281.1 - 5 480
BC048674.1 - 6 758 6774125

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-x666z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline