siRNA Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111111.1 | NP_001104581.1 | 5 | 691 | ||
| GenBank mRNA | AB476647.1 | - | 5 | 630 | ||
| AB476648.1 | - | 5 | 630 | |||
| AK155125.1 | - | 5 | 681 | |||
| AK157410.1 | - | 5 | 670 | |||
| AK158186.1 | - | 5 | 631 | |||
| BC060054.1 | - | 5 | 854 | |||

Atg16l2
Genomic Map