siRNA Details
| Gene Symbol: 4933427G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028955.1 | NP_083231.1 | 3, 4 | 535 | ||
| GenBank mRNA | AK016951.1 | - | 3, 4 | 535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933427G17Rik| Gene Symbol: 4933427G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028955.1 | NP_083231.1 | 3, 4 | 535 | ||
| GenBank mRNA | AK016951.1 | - | 3, 4 | 535 | ||