This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4930519F16RIK SIRNA Assays ›
Gene Symbol:
4930519F16Rik
Gene Name:
RIKEN cDNA 4930519F16 gene
Gene Aliases:
Ppnx
Chromosome Location:
Chr.X: 103182991 - 103256632 on Build GRCm38
UniGene ID:
Mm.1441
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 508516

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930519F16Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029170.1 NP_083446.1 4 999
GenBank mRNA AK039711.1 - 4 999
BC107248.2 - 4 1001 40052342
BC107249.1 - 4 997 40052344
BC114984.1 - 4 1001 40052347
BC115753.1 - 4 999 40052345

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-k8jnn:80/100.66.77.200:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0